Grey Coated Tyvek® UV Light Blocking GMP Equipment Covers
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
COVGTVK363235Z COVGTVK363268Z COVGTVK563666Z COV2PGTVK60445 COVGTVK604860Z COVGTVK563852Z
129-2900EA 52820 DKK
129-2900 129-2901 129-2902 129-2898 129-2833 129-2867
Grey Coated Tyvek® UV Light Blocking GMP Equipment Covers
Covers
Grey Tyvek® GMP covers block UV light, which can damage light-sensitive biotech products. Use in any pharmaceutical manufacturing setting to keep cleaned equipment protected from contamination.
  • Manufactured and packaged in a controlled cleanroom environment
  • Blocks UV light
  • Reduces labor costs
  • Simplifies the conformance to SOP's
  • Tear- and puncture-resistant
  • Hydrophobic
  • Lint-free
Grey UV Light blocking Tyvek® Covers can be used in all pharmaceutical manufacturing environments to help minimize microbial and particulate contamination risks.
Certificeringer: Certificates of conformance included.
Emballering: Covers are individually poly bagged, packaged 1 per labeled poly bag with lot number and DOM.