Ready-to-Load™ kits, mapping of restriction sites on plasmid DNA
Leverandør: EDVOTEK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
105 105-C
EDVO105EA 887 DKK
EDVO105 EDVO105-C
Ready-to-Load™ kits, mapping of restriction sites on plasmid DNA
Undervisningsmateriale Biologisk undervisningsmateriale Genetics Learning Activities
DNA mapping is a common procedure used to determine the location of genes. In this experiment, DNA markers and pre-digested plasmid DNA fragments are mapped using agarose gel electrophoresis.
  • For six lab groups
Also required: Electrophoresis tank, power supply, micropipettes (5 to 50 µl adjustable or 40 µl fixed volume) with tips, and a white light box is recommended.
Leveringsinformation: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution, electrophoresis buffer, InstaStain® DNA stain, 1 ml calibrated drop pipette, 100 ml graduated cylinder and microtipped transfer pipettes.