Ready-to-Load™ kits, DNA fingerprinting I: ID of DNA by restriction fragmentation patterns
Leverandør: EDVOTEK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
109 109-C
EDVO109EA 887 DKK
EDVO109 EDVO109-C
Ready-to-Load™ kits, DNA fingerprinting I: ID of DNA by restriction fragmentation patterns
Undervisningsmateriale Retsmedicinsk undervisningsmateriale Retsmedicinske sæt
Basic concepts of DNA fingerprinting are featured in this Ready-to-Load™ kit by comparing crime scene DNA with suspect DNAs. Fingerprint patterns are separated by agarose gel electrophoresis and the students determine who may have commited the crime.
  • For 6 lab groups
Also required: Electrophoresis tank, power supply, micropipettes (5 to 50 µl adjustable or 40 µl fixed volume) with tips, and a white light box is recommended.
Leveringsinformation: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution, electrophoresis buffer, InstaStain® DNA stain, 1 ml calibrated drop pipette, 100 ml graduated cyclinder and microtipped transfer pipettes.