Ready-to-Load™ kits, analysis of Eco RI cleavage patterns of lambda DNA
Leverandør: EDVOTEK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
112 112-C
EDVO112EA 887 DKK
EDVO112 EDVO112-C
Ready-to-Load™ kits, analysis of Eco RI cleavage patterns of lambda DNA
Undervisningsmateriale Biologisk undervisningsmateriale Genetics Learning Activities
Ready-to-Load™ kit for introducing the use of restriction enzymes as a tool to digest lambda DNA at specific nucleotide sequences.
  • For 6 lab groups
Also required: Electrophoresis tank, power supply, micropipettes (5 to 50 µl adjustable or 40 µl fixed volume) with tips, and a white light box is recommended.
Leveringsinformation: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution, electrophoresis buffer, InstaStain® DNA stain, 1 ml calibrated drop pipette, 100 ml graduated cyclinder and microtipped transfer pipettes.