Single channel pipettes, fixed volume, MiniFIX
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1285 1286 1287 1288 1289 1290 1291 1292 1293 1294
613-1391EA 313 DKK
613-1391 613-1392 613-1393 613-1394 613-1395 613-1396 613-1397 KART1292 KART1293 KART1294
Single channel pipettes, fixed volume, MiniFIX
Pipetter Automatiske pipetter
Compact, light weight (10 g) pipettes - an accurate, but low cost alternative to plastic Pasteur pipettes.
  • Colour coded button for easy volume identification
  • Fits Kartell and all other common pipette tips
Leveringsinformation: Supplied in blister pack with calibration and performance certification.