Horizontal Electrophoresis Apparatus, M6 and M12
Leverandør: EDVOTEK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
500 502 504
732-1417EA 2770 DKK
732-1417 732-1419 732-1420
Horizontal Electrophoresis Apparatus, M6 and M12
Elektroforesesystemer Horisontale Gelelektroforesesystemer
Capable of running one or two groups of student samples at the same time and producing excellent results in 30 to 40 minutes.
  • Seamless, injection-moulded casing
  • Safety interlock cover
  • Corrosion-resistant platinum electrodes
  • Safety insulated electrical leads
  • Adjustable leveling feet