Atlas® SPE Plates, 96-well, RCH
Leverandør: Tecan
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30163872 30163864 30163033 30160524
TCAT30163872EA 0 DKK
TCAT30163872 TCAT30163864 TCAT30163033 TCAT30160524
Atlas® SPE Plates, 96-well, RCH
Mikroplader Solid Phase Ekstraktions plader
A hydrophilic/lipophilic balanced chemistry offering selectivity to a wide range of analytes. It provides enhanced selectivity for polar species, also comes in larger particle for more viscous samples. A direct alternative to Waters Oasis® HLB and Phenomenex Strata X.
  • Resin chemistry: N-vinylpyrrolidone-divinylbenzene copolymer
  • Particle shape: spherical
  • pH stability: 1 - 14
Ideal for use in small molecules and pharmaceutical drugs, sample preparation for drug monitoring, drug screening, and steroid analysis workflows.