CRS ZPure™ H₂O Gas Filters
Leverandør: Trajan Scientific and Medical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
202230-B 202230D-B 202230D-SS 202230L-B 202230L-SS 202230-SS 202230XL-B 202230XL-SS 202231-B 202231D-B 202231D-SS 202231L-B 202231L-SS 202231-SS 202231XL-B 202231XL-SS 203011-B 203011L-B 203011L-SS 205705 205706 205707 202230-4-PC 202230-6-PC 202230-B-QC 202230D-4-PC 202230D-6-PC 202230D-B-QC 202230D-SS-QC 202230L-4-PC 202230L-6-PC 202230L-B-QC 202230L-SS-QC 202230-SS-QC 202230XL-4-PC 202230XL-6-PC 202230XL-B-QC 202230XL-SS-QC 202231-B-QC 202231D-B-QC 202231D-SS-QC 202231L-B-QC 202231L-SS-QC 202231-SS-QC 202231XL-B-QC 202231XL-SS-QC 202269XL-SS 202RH2-B-QC 202RH2D-B-QC 202RH2D-SS-QC 202RH2L-B-QC 202RH2L-SS-QC 202RH2-SS-QC 202RH2XL-B-QC 202RH2XL-SS-QC
SGEA202230-BEA 1270 DKK
SGEA202230-B SGEA202230D-B SGEA202230D-SS SGEA202230L-B SGEA202230L-SS SGEA202230-SS SGEA202230XL-B SGEA202230XL-SS SGEA202231-B SGEA202231D-B SGEA202231D-SS SGEA202231L-B SGEA202231L-SS SGEA202231-SS SGEA202231XL-B SGEA202231XL-SS SGEA203011-B SGEA203011L-B SGEA203011L-SS SGEA205705 SGEA205706 SGEA205707 SGEA202230-4-PC SGEA202230-6-PC SGEA202230-B-QC SGEA202230D-4-PC SGEA202230D-6-PC SGEA202230D-B-QC SGEA202230D-SS-QC SGEA202230L-4-PC SGEA202230L-6-PC SGEA202230L-B-QC SGEA202230L-SS-QC SGEA202230-SS-QC SGEA202230XL-4-PC SGEA202230XL-6-PC SGEA202230XL-B-QC SGEA202230XL-SS-QC SGEA202231-B-QC SGEA202231D-B-QC SGEA202231D-SS-QC SGEA202231L-B-QC SGEA202231L-SS-QC SGEA202231-SS-QC SGEA202231XL-B-QC SGEA202231XL-SS-QC SGEA202269XL-SS SGEA202RH2-B-QC SGEA202RH2D-B-QC SGEA202RH2D-SS-QC SGEA202RH2L-B-QC SGEA202RH2L-SS-QC SGEA202RH2-SS-QC SGEA202RH2XL-B-QC SGEA202RH2XL-SS-QC
CRS ZPure™ H₂O Gas Filters
Filtre Gasfiltre
The ZPure™ line of filters removing a wide range of contaminants to trace levels.
  • High efficiency in-line traps with outstanding capacity
  • High quality activated adsorbents for long purifier life and efficient contaminant removal
  • various size and fitting configurations to fit existing installations
  • filter lifetime is dependent on quality of incoming gas and the flow rate
  • Individually leak tested
It can be used to remove from inert gases, HE, AR, N₂, H₂ methane and clean dry air (CDA) to low ppb levels. It is recommended for any application requiring ultra dry gas.