Cerex® Atlas® SCX SPE Columns
Leverandør: Tecan
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30174702 30174710 30174714 30174718
TCAT30174702EA 0 DKK
TCAT30174702 TCAT30174710 TCAT30174714 TCAT30174718
Cerex® Atlas® SCX SPE Columns
Kromatografikolonner
A hydrophilic/lipophilic balanced strong cationic chemistry offering excellent selectivity to basic and neutral compounds. A direct alternative to Waters Oasis® MCX and Phenomenex Strata XC.
  • Mode: Ion-exchange
  • Liquid handler compatible
Atlas® is a hydrophilic/lipophilic balanced polymeric sorbent with a N-vinylpyrrolidone-divinyl-ben-zene copolymer. Atlas® SCX which incorporates the Atlas’s copolymer technology along with a strong cationic exchange chemistry to provide a focused clean up when working with basic compounds in tough matrices.