Cerex® TRACE-B SPE Columns
Leverandør: Tecan
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
711-0101R 711-0201R 711-0506C 711-335C
TCAT711-0101REA 0 DKK
TCAT711-0101R TCAT711-0201R TCAT711-0506C TCAT711-335C
Cerex® TRACE-B SPE Columns
Kromatografikolonner
Strong cationic exchange chemistry ideal for aggressive cleanup of basic target compounds.
  • Ion-exchange
  • Liquid handler compatible
A 15 µm polymer micro particulate with strong cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Ideal for basic target compounds. Formulated for pain panel/FNT.