jo_Clean Strap Mid, S3 ESD Safety Shoes, White
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
16681/48 16681/36 16681/37 16681/38 16681/39 16681/40 16681/41 16681/42 16681/43 16681/44 16681/45 16681/46 16681/47
ELTN16681/48EA 440 DKK
ELTN16681/48 ELTN16681/36 ELTN16681/37 ELTN16681/38 ELTN16681/39 ELTN16681/40 ELTN16681/41 ELTN16681/42 ELTN16681/43 ELTN16681/44 ELTN16681/45 ELTN16681/46 ELTN16681/47
jo_Clean Strap Mid, S3 ESD Safety Shoes, White
Sko og støvler
Thanks to its excellent discharge capability, the shoe is suitable for
work in ESD sensitive or electrostatically protected areas (EPA).
  • Full, padded bellows tongue
  • MONO-PU sole jo_MONO
  • Composite toe cap
  • Metal- and leather-free equipment
  • Available in white finish
Hydrophobised microfibre
Fabric lining for natural ventilation. Full-length inlay sole. Non-metallic, penetration-resistant midsole.
Certificeringer: EN ISO 20345 S3 SRC, form B, ESD Standard, The shoes comply to the standard 61340-5-1, Certification in accordance with DGUV rule 112-191.