Aggressive Adhesive Self-Laminating Vinyl Wrap-Around Labels with Ribbon for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
170957 170958 170959 170960 170961 170962 170963 170964
BRDY170957EA 487 DKK
BRDY170957 BRDY170958 BRDY170959 BRDY170960 BRDY170961 BRDY170962 BRDY170963 BRDY170964
Aggressive Adhesive Self-Laminating Vinyl Wrap-Around Labels with Ribbon for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketter
B-417 High-adhesion self-laminating vinyl wraps are designed for tough conditions, including high-humidity environments, small diameter wires and newer wire jacketing materials.
  • Dependable, long-term adhesion to small diameter wires
  • Will stay adhered in even the most humid environments
  • Strong adhesion to a variety of wire jacketing materials, including Teflon™, silicone, braided and traditional PVC
  • Features a white printable area and a clear overlaminating area that protects the print
  • Ideal for control panels, electrical panels, wire harnesses, datacom, and wire and cable ID
  • Matte finish
Ideal for data and telecommunications labelling, electrical labelling and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction and better performance and reliability. A ribbon is required to print this material. Use part number M5-116-417.
Matte finish, chemical-resistant, humidity-resistant, oil-resistant, water-resistant, weather-resistant, indoor/outdoor use B-417 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511.
Certificeringer: Compliant with directive 2011/65/EU and 2015/863.