Magnetic Polyester Label Tape for M710 Printer
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
173985 173986
BRDY173985EA 553 DKK
BRDY173985 BRDY173986
Magnetic Polyester Label Tape for M710 Printer
Etiketter
These magnetic labels used for safety and facility identification. They offer extra flexibility for lean/5S initiatives as they can be easily repositioned or reused.
  • Non-adhesive magnetic material can be thermal transfer printed
  • Offers an average outdoor durability of 3 to 5 years
  • Labels withstand moderate heat, mild chemicals, oil and water
  • Service temperature of −18 to +70 °C
  • Brady Authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability
Certificeringer: Compliant with Directive 2011/65/EU and 2015/863.