ReproGel Gel Permeation Chromatography
Leverandør: DR MAISCH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RG5.500.S3020 RG5.500.S6020
MAISRG5.500.S3020EA 26510 DKK
MAISRG5.500.S3020 MAISRG5.500.S6020
ReproGel Gel Permeation Chromatography
Kromatografikolonner
Reprogel GPC is a polymer based me for non-aqueous gel permeation chromatography of polymers. A range of 7 phases with different pore sizes separate polymers with a MW ranging from <1000 Da up to 10000 kDa. Column are used under 100% THF.
  • Used to determine the relative molecular weight of polymer samples as well as the distribution of molecular weights