PermaSleeve® Low Smoke Zero Halogen Wire Marking Sleeves for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
170848 170849 170951 170952 170953 170954 170955 170956
BRDY170848EA 853 DKK
BRDY170848 BRDY170849 BRDY170951 BRDY170952 BRDY170953 BRDY170954 BRDY170955 BRDY170956
PermaSleeve® Low Smoke Zero Halogen Wire Marking Sleeves for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketter
Permanently mark wires and cables with the low-smoke, halogen-free B-7641 polyolefin label material. Pre-sized to fit most wire and cable assemblies. Print with a Brady handheld label printer.
  • Halogen free
  • Chemical resistant
  • Fuel and oil resistant
  • Heat resistant
  • Low temperature resistant
  • Specialised - Designed for mass-transit and marine applications where flame-retardant, low-smoke wire marking materials are required
  • Durable - Flame-retardant and resistant to temperatures from –30 to +105 °C
  • Efficient - Fast shrinking with a 2:1 shrink ratio; permanently adheres to wires quickly with a heat gun (Brady item PMS-HG-1)
Matte finish, chemical-resistant, corrosion-resistant, humidity-resistant, self-extinguishing, UV-resistant, weather-resistant B-7641 labels are compatible with Brady printer models and M410, BMP41, M510, BMP51, BMP53, M511.
Certificeringer: Compliant with Directive 2011/65/EU and 2015/863; BS-6853 (1999) Vehicle Category 1A; EN-45545-2, requirement 24, Vehicle Category HL3, Zero Halogen.