Continuous Repositionable Vinyl Cloth Terminal Block Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
170785 170782 170783 170784 170787 170788 170789 170790 170791 170792 170926 M4-49-498
BRDY170785EA 348 DKK
BRDY170785 BRDY170782 BRDY170783 BRDY170784 BRDY170787 BRDY170788 BRDY170789 BRDY170790 BRDY170791 BRDY170792 BRDY170926 BRDYM4-49-498
Continuous Repositionable Vinyl Cloth Terminal Block Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketter
Easily place, remove and reposition labels on any clean, dry surface with the B-498 CleanLift® series vinyl cloth label material. Print almost anywhere with a high-quality Brady handheld label printer.
  • Repositionable
  • Heat resistant
  • Low temperature resistant
  • Abrasion resistant
  • Semi-gloss finish
Ideal for marking wires, cables, terminal blocks, wall plates. Constructed of tough coated vinyl cloth for resistance to extreme temperatures. Made with a strong rubber-based adhesive that can be cleanly removed and securely replaced on any clean, dry surface.
Semi-gloss finish, abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant B-498 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511.
Certificeringer: UL recognised to UL969, RoHS Compliant to 2005/618/EC MCV.