PIG® Quick-Response High-Visibility Spill Carts
Leverandør: NEW PIG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KIT2401-999 KIT3401-999
NEWPKIT2401-999EA 7450 DKK
NEWPKIT2401-999 NEWPKIT3401-999
PIG® Quick-Response High-Visibility Spill Carts
Spild materialer
Get to the spill fast and fully stocked with easy-to-access absorbents in this mobile, high-visibility yellow spill kit.
  • Absorb anything from water or coolants to oil or solvents with the pre-packed absorbents inside this spill caddy
  • Manoeuvre effortlessly, even through doorways, thanks to the tip-back design and snag-resistant 18 cm wheels
  • Contents are compartmentalised for easy access
Easily work to clean any small- or moderate-sized spill since the spill caddy's spacious interior holds 75 L worth of absorbents and spill-response items, and the lockable front-loading doors open wide for convenient access; lock with a padlock (not included) to prevent pilferage.
Keep everyone alert with the high-visibility yellow colour that makes the caddy easy to spot and grab, and provides visibility for others when spill-response is in action; store anywhere indoors.
Advarsel: HAZ-MAT/chemical notice: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase. If you have any questions or need samples to test, please call technical services.