Tanks for water baths
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
43187003 43194001
HECH43187003EA 1440 DKK
HECH43187003 HECH43194001
Tanks for water baths
Tanke Firkantede beholdere
Tubs made of Plexiglas®, available with or without sloping base. Sloping base facilitates circulation of the liquid.
For use with water bath thermostats.