Showa 7565 Nitrile Gloves
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7565L 7565M 7565S 7565XL 7565XXL
BEST7565LEA 343 DKK
BEST7565L BEST7565M BEST7565S BEST7565XL BEST7565XXL
Showa 7565 Nitrile Gloves
Handsker
These powder-free disposable gloves are made of 100% nitrile. They provide protection against chemical penetration and projection.
  • Antistatic
  • Chemical-resistant, water-resistant, increased sensitivity
  • Forearm protection
  • Latex, silicone, plasticisers-free
  • Reduce irritation and allergy risks
The single-use glove is chlorinated to enhance comfort and reduce tackiness within the glove. An added textured finish increased grip without reducing fingertip sensitivity.
Typical applications include: Aerospace, electronics, food, pharmaceutical, healthcare, automative, and laboratory.
Certificeringer: EN ISO 374-1:2016/Type B; EN ISO 374-5:2016; Cat III.
Food contact approved.