Self-Sealing Sterilisation Peel Pouches, Medical Paper/Film, with Steam Indicator
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PPSS035X09CF PPSS055X105 PPSS075X13CF PPSS08X16CF PPSS10X14CF PPSS035X055CF PPSS035X09 PPSS075X13 PPSS12X18 PPSS12X18CF PPSS035X055 PPSS055X105CF PPSS08X16 PPSS10X14 PPSS13X18CF PPSS15X20CF PPSS18X22CF
KACCPPSS035X09CFEA 1360 DKK
KACCPPSS035X09CF KACCPPSS055X105 KACCPPSS075X13CF KACCPPSS08X16CF KACCPPSS10X14CF KACCPPSS035X055CF KACCPPSS035X09 KACCPPSS075X13 KACCPPSS12X18 KACCPPSS12X18CF KACCPPSS035X055 KACCPPSS055X105CF KACCPPSS08X16 KACCPPSS10X14 KACCPPSS13X18CF KACCPPSS15X20CF KACCPPSS18X22CF
Self-Sealing Sterilisation Peel Pouches, Medical Paper/Film, with Steam Indicator
Poser Steriliseringsposer
Designed for steam sterilisation cycles (121 to 135 °C).
  • Peels open cleanly
  • Triple weld factory seals
  • Convenient, time-saving self-seal closure system
  • Strong, rip- and tear-resistant 60 g/m² medical grade paper material
  • Good barrier characteristics prevent microbial penetration
  • Sub-packed into sealed labelled poly bags with manufacturing lot number
  • Date of manufacture on label for complete traceability
Significant improvement versus cutting and wrapping with sheets and autoclave tape.
Certificeringer: Certificates of Conformance sent with each shipment.