Self-Laminating Cryogenic Polyester Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
170882 170877 170767 170765 170875 170766 170768 170769 170878 170879 170880 170881 170883
BRDY170882EA 562 DKK
BRDY170882 BRDY170877 BRDY170767 BRDY170765 BRDY170875 BRDY170766 BRDY170768 BRDY170769 BRDY170878 BRDY170879 BRDY170880 BRDY170881 BRDY170883
Self-Laminating Cryogenic Polyester Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketter
Securely label laboratory tubes and vials for cryogenic storage with this self-laminating, ultra-thin polyester label material (B-461). Print almost anywhere with a compact Brady portable label printer.
  • For use on smooth, curved surfaces
  • Withstands extreme temperatures from −196 to +130 °C
  • Chemical-resistant
  • Transparent overlaminate protects print
Versatile labels are ideal for curved surfaces such as vials, centrifuge tubes, test tubes, straws, and slides. Self-Laminating Cryogenic Polyester Labels are able to withstand extreme temperatures from −196 to +130 °C , immersion in liquid nitrogen, and autoclave use. Resistant to ethanol, toluene, isopropanol, xylene, DMSO, 50% acetic acid, 10% hydrochloric acid, and 10% Clorox solution. A transparent overlaminate protects the print and maintains content visibility. For material specific performance and properties, please refer to the Technical Data Sheet (TDS).
Matte finish, chemical-resistant, low and high temperature-resistant, smear-resistant B-461 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511.
Certificeringer: RoHS compliant to 2011/65/EU (Pass 2015/863).
Emballering: Labels are packaged 180 per box.