Biohazard Bags, Without Print, Autoclavable, Sterile
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3138CBHNPST10D 3138CBHNPST10S
KACC3138CBHNPST10DEA 3940 DKK
KACC3138CBHNPST10D KACC3138CBHNPST10S
Biohazard Bags, Without Print, Autoclavable, Sterile
Poser Affaldsposer
Pre-sterilised and packaged in multiple layers of Tyvek peel pouches and poly bags to gain access into the aseptic area without excessive spraying and wiping with sanitisation agents. The bags are used for the safe and effective removal of biohazardous materials from the aseptic area through the decontamination autoclave.
  • Manufactured and packaged in an ISO certified cleanroom
  • Tear and puncture resistant 2 mil clear polypropylene
  • Ten bags are double packaged in Tyvek/film chevron peel pouches and then inside a third, sealed poly bag layer
  • Labelled with lot number, date of manufacture and expiration date
  • Autoclavable at 135 °C (or 275 °F) for 15 minutes
  • Validated sterile to a 10⁻⁶ SAL
  • Certificates of conformance and sterility sent with each shipment
Certificeringer: CoC and certificate of sterility.