VWR® HDPE Bottles, Narrow Neck
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
215-4539EA 177 DKK
215-4539 215-4540 215-4541
VWR® HDPE Bottles, Narrow Neck
Flasker Plastflasker
These bottles are made with 100% virgin HDPE resin.
  • Leak proof sealing
Narrow mouth bottle ideal for liquid storage.
Certificeringer: Compliance to US FDA 21 CFR and USP Class VI.