Hi-Vis 3-in-1 Contrast Lite Jackets
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S162ONRL S162ONRM S162ONRS S162ONRXL S162ONRXXL S162ONRXXXL
PRTWS162ONRLEA 726 DKK
PRTWS162ONRL PRTWS162ONRM PRTWS162ONRS PRTWS162ONRXL PRTWS162ONRXXL PRTWS162ONRXXXL
Hi-Vis 3-in-1 Contrast Lite Jackets
Arbejdstøj
This 3-in-1 addition to the Hi-Vis 150D Oxford range is excellent value for money and provides a combination of supreme versatility with innovative design. Complete with multi-layered practical pockets, full front zip closure, concealed hood and a detachable stand-alone internal fleece jackets.
  • Waterproof with taped seams preventing water penetration
  • Extremely water resistant eco-conscious fabric finish, water beads away from fabric surface
  • Detachable inner jackets for added versatility in all weather conditions
  • Pack away hood for added functionality
  • Stud adjustable cuffs for a secure fit
  • Contrast panels for protection against dirt
Storm flap front to protect against the elements. ID card holder for ID card display. Reflective tape for increased visibility. Jackets can be worn three ways for multi use. With nine pockets for ample storage. Radio loop for easy clipping of a radio. Detachable lining for added versatility in all weather conditions. 40+ UPF rated fabric to block 98% of UV rays. Waterproof with taped seams preventing water penetration.
Weather conditions: Dark: High visibility, reflective clothing
Certificeringer: EN ISO 20471 Class 3
RIS 3279 TOM Issue 2 (Orange Only)
EN 343 Class 3:1 X WP 5000 mm