FreezerBondz™ Ultra-Thin Polyester Laboratory Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
170781 170897 170898 170892 170901 170777 170893 170895 170896 170900
BRDY170781EA 776 DKK
BRDY170781 BRDY170897 BRDY170898 BRDY170892 BRDY170901 BRDY170777 BRDY170893 BRDY170895 BRDY170896 BRDY170900
FreezerBondz™ Ultra-Thin Polyester Laboratory Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketter
Label curved frozen lab surfaces including tubes, vials and cryocanes with the ultra-thin B-492 polyester label material. Print almost anywhere with a durable Brady handheld label printer.
  • Ideal for laboratory use on curved surfaces including test tubes, vials, and straw pipettes
  • Withstands chemicals, abrasions, and extreme temperatures, including storage in liquid nitrogen
  • Made with a permanent acrylic adhesive for tight bonding onto smooth curved surfaces such as glass and polypropylene
  • Aggressive, permanent adhesive
  • Matte finish
Matte finish, chemical-resistant, smear-resistant B-492 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511. Perfect for general identification and laboratory labelling, these labels should be used with the ribbon series R6400.
Certificeringer: RoHS compliant to 2011/65/EU (Pass 2015/863).
Emballering: Labels are packaged 240 per box.