Compositelite™ Protector S3 HRO ESD Safety Boots, Lace-Up
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FD37BKB39 FD37BKB40 FD37BKB41 FD37BKB42 FD37BKB43 FD37BKB44 FD37BKB45 FD37BKB46 FD37BKB47
PRTWFD37BKB39EA 672 DKK
PRTWFD37BKB39 PRTWFD37BKB40 PRTWFD37BKB41 PRTWFD37BKB42 PRTWFD37BKB43 PRTWFD37BKB44 PRTWFD37BKB45 PRTWFD37BKB46 PRTWFD37BKB47
Compositelite™ Protector S3 HRO ESD Safety Boots, Lace-Up
Sko og støvler
The boot height ensures the ankle is protected and supported, with an angled top line for better fit and leg flexibility. A padded collar offers exceptional comfort.
  • Suitable for use in ESD environments
  • Heat-resistant outsole to 300 °C
  • Enhanced tread pattern for a better traction on uneven terrain
  • Padded collar for added comfort
  • Protective scuff cap and composite toe cap
  • Pierce-resistant composite midsole
  • Anti-static footwear
  • Energy absorbing seat region
  • Fuel and oil resistant outsole
  • 100% non-metallic
  • SRC - Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Dual density sole unit
The Compositelite protector safety boot features a heat-resistant outsole, ESD properties, and excellent slip resistance. The engineered tread pattern aids agility on uneven terrain while also being resistant to fuel oil and water. A bellow tongue construction provides better resistance to water penetration, along with a water-resistant upper to prevent water penetration.
Certificeringer: EN ISO 20345: 2011 S3. IEC 61340-5-1:2016. CE certified.