Powdered Latex Disposable Gloves
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A910WHRM A910WHRXL
PRTWA910WHRMEA 66 DKK
PRTWA910WHRM PRTWA910WHRXL
Powdered Latex Disposable Gloves
Handsker
Manufactured from genuine natural rubber (which may cause allergic reactions in some individuals), these gloves combine the highest degree of dexterity available with superior strength and comfort.
  • Ambidextrous design suitable for left- and right-handed users
  • Acceptable quality limit (AQL) 1.5
  • 240 mm length
  • 100 gloves per box
  • This product is sold in inner boxes
Super soft for superior user comfort and super stretchy to reduce hand fatigue, these gloves are smooth and non-textured with a beaded cuff. Retail box aids presentation for retail sales.
Colour: White
Certificeringer: EN ISO 21420: 2020 Dexterity 5. CE Cat 1. CE certified.