Accessories for Endotoxin Tests
Leverandør: FUJIFILM WAKO CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LRW-12125 LRW-2030
WAKFLRW-12125EA 3280 DKK
WAKFLRW-12125 WAKFLRW-2030
Accessories for Endotoxin Tests
Mikrobiologiske testsystemer Endotoxin test
LAL Reagent Water can be used to fulfill all aspects of endotoxin testing. From reconstitution of both the CSE and LAL for the creation of standard curves to the making of product dilutions, LAL reagent water provides reliable and reproducible data when paired with our endotoxin-specific LAL reagents.
  • Endotoxin-free water (<0,001 EU/ml)
  • Great for reconstitution of LAL, RSE and CSE
  • Used to perform dilution series of RSE and CSE
  • Used for sample dilutions