ISAspher HPLC Columns, Silica
Leverandør: ISERA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A111-423A00 A111-423B00 A111-423C00 A111-423D00 A111-423E00 A111-423F00 A111-423G00 A111-423H00 A111-423J00 A111-423K00 A111-423L00 A111-423M00 A111-423N00 A111-423P00 A111-423Q00 A111-423R00 A111-423S00 A111-423T00 A111-423U00 A111-423V00 A111-423W00 A111-423X00 A111-423Y00 A111-423Z00 A111-425A00 A111-425B00 A111-425C00 A111-425D00 A111-425E00 A111-425F00 A111-425G00 A111-425H00 A111-425J00 A111-425K00 A111-425L00 A111-425M00 A111-425N00 A111-425P00 A111-425Q00 A111-425R00 A111-425S00 A111-425T00 A111-425U00 A111-425V00 A111-425W00 A111-425X00 A111-425Y00 A111-425Z00 A111-429A00 A111-429B00 A111-429C00 A111-429D00 A111-429E00 A111-429F00 A111-429G00 A111-429H00 A111-429J00 A111-429K00 A111-429L00 A111-429M00 A111-429N00 A111-429P00 A111-429Q00 A111-429R00 A111-429S00 A111-429T00 A111-429U00 A111-429V00 A111-429W00 A111-429X00 A111-429Y00 A111-429Z00
ISERA111-423A00EA 2160 DKK
ISERA111-423A00 ISERA111-423B00 ISERA111-423C00 ISERA111-423D00 ISERA111-423E00 ISERA111-423F00 ISERA111-423G00 ISERA111-423H00 ISERA111-423J00 ISERA111-423K00 ISERA111-423L00 ISERA111-423M00 ISERA111-423N00 ISERA111-423P00 ISERA111-423Q00 ISERA111-423R00 ISERA111-423S00 ISERA111-423T00 ISERA111-423U00 ISERA111-423V00 ISERA111-423W00 ISERA111-423X00 ISERA111-423Y00 ISERA111-423Z00 ISERA111-425A00 ISERA111-425B00 ISERA111-425C00 ISERA111-425D00 ISERA111-425E00 ISERA111-425F00 ISERA111-425G00 ISERA111-425H00 ISERA111-425J00 ISERA111-425K00 ISERA111-425L00 ISERA111-425M00 ISERA111-425N00 ISERA111-425P00 ISERA111-425Q00 ISERA111-425R00 ISERA111-425S00 ISERA111-425T00 ISERA111-425U00 ISERA111-425V00 ISERA111-425W00 ISERA111-425X00 ISERA111-425Y00 ISERA111-425Z00 ISERA111-429A00 ISERA111-429B00 ISERA111-429C00 ISERA111-429D00 ISERA111-429E00 ISERA111-429F00 ISERA111-429G00 ISERA111-429H00 ISERA111-429J00 ISERA111-429K00 ISERA111-429L00 ISERA111-429M00 ISERA111-429N00 ISERA111-429P00 ISERA111-429Q00 ISERA111-429R00 ISERA111-429S00 ISERA111-429T00 ISERA111-429U00 ISERA111-429V00 ISERA111-429W00 ISERA111-429X00 ISERA111-429Y00 ISERA111-429Z00
ISAspher HPLC Columns, Silica
Kromatografikolonner
HPLC column of the ISAspher line guarantee the highest possible reliability in a wide range of applications.
  • The production of the silica used as well as the functionalisation are carried out according to highly developed and validated process
  • Reliable and proven
  • Flexibility and robustness for every HPLC application
This ensure unchanged properties of the stationary phases in terms of selectivity and consistent chemical properties.
Direct scale-up e.g. starting from a particle size of 5 µm to semi-preparative or preparative applications (up to 45 µm particle size) can be easily implemented. Of course, the same applies for a scale-down, e.g. to a 3 µm particle size, if a speed-optimised analytical method has to be established.
Certificeringer: Individual test certificate is enclosed with each column.
Leveringsinformation: Columns are delivered in solvent which is suitable for normal phase chromatography.