Destructible Tamper Evident Matte Vinyl Labels for M710 and BMP71
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
174601 173413 174608 173386
BRDY174601EA 420 DKK
BRDY174601 BRDY173413 BRDY174608 BRDY173386
Destructible Tamper Evident Matte Vinyl Labels for M710 and BMP71
Etiketter
Tamper-resistant vinyl (B-351) is designed to fracture easily in order to show signs of product tampering and to prevent on-piece label removal.
  • High performance white matte vinyl with permanent adhesive
  • Ideal for applications that require resistance to product tampering
  • Use caution when removing from liner as material is fragile
  • Service temperature of −40 to +82 °C (−40 to +180 °F)
Brady Authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6200.