Chemical and Temperature Resistant Polypropylene Labels for M610, M611 and BMP61
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
174115 174101 174078 173497 173513 173508 173495
BRDY174115EA 1020 DKK
BRDY174115 BRDY174101 BRDY174078 BRDY173497 BRDY173513 BRDY173508 BRDY173495
Chemical and Temperature Resistant Polypropylene Labels for M610, M611 and BMP61
Etiketter
Polypropylene (B-7425) material is ideal for general purpose barcode applications and withstands liquid nitrogen, autoclaving, and other harsh environments.
  • Matte white polypropylene with acrylic adhesive is thermal transfer printable
  • Perfect for laboratory identification such as vials, centrifuge tubes, and test tubes
  • Service temperature of −80 to +70 °C (−112 to +158 °F)
Ideal for component and equipment labelling, general identification, laboratory labelling, and temperature indicating labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310.