Aggressive Adhesive Self-Laminating Vinyl Wrap Around Labels for M710 Printer
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
170485 170489 170491 170490 170484 170483 170487 170488 170492 170486
BRDY170485EA 442 DKK
BRDY170485 BRDY170489 BRDY170491 BRDY170490 BRDY170484 BRDY170483 BRDY170487 BRDY170488 BRDY170492 BRDY170486
Aggressive Adhesive Self-Laminating Vinyl Wrap Around Labels for M710 Printer
Etiketter
B-417 High-adhesion self-laminating vinyl wraps are designed for tough conditions, including high-humidity environments, small diameter wires and newer wire jacketing materials.
  • Dependable, long-term adhesion to small diameter wires
  • Will stay adhered in even the most humid environments
  • Strong adhesion to a variety of wire jacketing materials, including Teflon™, silicone, braided and traditional PVC
  • Features a white printable area and a clear overlaminating area that protects the print
  • Ideal for control panels, electrical panels, wire harnesses, datacom, and wire and cable ID
  • Matte finish
Ideal for data and telecommunications labelling, electrical labelling and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction and better performance and reliability. A ribbon is required to print this material. Use part number M7-R4300.
Certificeringer: UL recognised to UL969 labelling and marking standard when printed with the Brady series R4300 and R7950 ribbons. See UL files MH62024 for specific details. Brady customers should specify at purchase if they require UL recognised B-417 labels. Brady sells both UL recognised B-417 labels and non-approved labels depending on customer requirements.