Multi-Purpose Matte Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
174260 174269 174232
BRDY174260EA 551 DKK
BRDY174260 BRDY174269 BRDY174232
Multi-Purpose Matte Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Etiketter
B-459 Polyester from Brady® is designed for applications such as topside of printed circuit boards and rating plates that use high quality barcodes and graphics.
  • Matte white polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Excellent resistance to most chemicals, oils, and water
  • Service temperature of −40 to 100 °C (−40 to 212 °F)
Ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, general identification, and inventory and inspection labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R6000 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certificeringer: UL recognised.