Ultra Aggressive Adhesive Multi-Purpose Matte Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
174263 174235 174297 174303 174284 173687 173881 173817 173719 173743 173684 173730 173828
BRDY174263EA 622 DKK
BRDY174263 BRDY174235 BRDY174297 BRDY174303 BRDY174284 BRDY173687 BRDY173881 BRDY173817 BRDY173719 BRDY173743 BRDY173684 BRDY173730 BRDY173828
Ultra Aggressive Adhesive Multi-Purpose Matte Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Etiketter
Ultra Aggressive Polyester (B-489A) with permanent adhesive is ideal for labelling products and general ID in both commercial and industrial applications.
  • Best for application on smooth and highly textured indoor surfaces, including metal and low surface energy plastics
  • Matte white polyester material has a permanent rubber based adhesive and is thermal transfer printable
  • Service temperature of −40 to 120 °C (−40 to 248 °F)
Ideal for asset tracking, barcode labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, and inventory and inspection labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certificeringer: UL recognised. CSA accepted.