Paper Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
173965 174293 174229 174241 173962 173872 173906
BRDY173965EA 436 DKK
BRDY173965 BRDY174293 BRDY174229 BRDY174241 BRDY173962 BRDY173872 BRDY173906
Paper Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Etiketter
Paper Labels with Permanent Rubber Adhesive (B-424) provide good contrast and smear resistance for thermal transfer and barcode printing applications.
  • White matte paper label stock features permanent rubber adhesive
  • Low cost is ideal for all general-purpose labelling projects
  • Service temperature of −40 to 50 °C (−40 to 122 °F)
Ideal for barcode labelling, electrical labelling, general identification, and laboratory labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.