Ultra-Thin Electrostatic Dissipative (ESD) Polyimide Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
173724 173711 173864 173895 173918 173792 173697
BRDY173724EA 945 DKK
BRDY173724 BRDY173711 BRDY173864 BRDY173895 BRDY173918 BRDY173792 BRDY173697
Ultra-Thin Electrostatic Dissipative (ESD) Polyimide Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Etiketter
Static Dissipative Polyimide (B-718) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labelling.
  • Low profile, high temperature, glossy white polyimide material
  • Static dissipative adhesive and static dissipative release liner
  • Surface resistivity values in the recommended range for Dissipative ESD packaging materials
  • Service temperature of −70 to 100 °C (−94 to 212 °F)
Ideal for circuit board labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R6000 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certificeringer: UL recognised. Meets the requirements of ANSI/ESD S541-2008 (between 10⁴ and 10¹¹ ohms) for dissipative ESD packaging materials. Meets the performance requirements of MIL-STD-202G, Method 215K.
Emballering: 750 label(s) per roll.