Mal-amido-PEG4-NHS ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2,5-Dioxopyrrolidin-1-yl 19-(2,5-dioxo-2H-pyrrol-1(5H)-yl)-17-oxo-4,7,10,13-tetraoxa-16-azanonadecan-1-oate, Mal-amido-PEG4-NHS ester, Maleimide-PEG4-succinimidyl Ester, Maleimide-PEG4-NHS Ester

M2185-250MG M2185-50MG
TCIAM2185-250MGEA 3530 DKK
TCIAM2185-250MG TCIAM2185-50MG
Mal-amido-PEG4-NHS ≥98.0% (by HPLC)
Mal-amido-PEG4-NHS
Formel: C₂₂H₃₁N₃O₁₁
Opbevaringstemperatur: Fryser
MDL Number: MFCD11041113
CAS-nummer: 756525-99-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR