Eppendorf twin.tec® Trace PCR Plates BioBased
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0030531.043 0030531.051 0030531.060 0030531.078 0030531.086
EPPE0030531.043EA 1120 DKK
EPPE0030531.043 EPPE0030531.051 EPPE0030531.060 EPPE0030531.078 EPPE0030531.086
Eppendorf twin.tec® Trace PCR Plates BioBased
Mikroplader PCR plader
Discover the PCR Plates BioBased made of bio-based polymer (mass balance approach). These plates allow you to significantly reduce your consumable-related carbon footprint and yet to maintain the identical technical performance. The new bio-based plates have the identical material properties and technical specifications as the legacy plates.
  • Reduction of consumable-related carbon footprint in the lab
  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
  • One-piece design: Combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean)
  • Available with barcode (upon request)
Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation.