VWR® Square Media Biotainer Canisters
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWRI210-000110EA 1620 DKK
VWRI210-000110 VWRI210-000107 VWRI210-000105 VWRI210-000108 VWRI210-000106 VWRI210-000109
VWR® Square Media Biotainer Canisters
Flasker Plastflasker
VWR® square containers are ideal for storage of media, laboratory reagents, and biological buffers. Eliminate washing, depyrogenation, and autoclaving steps with sterile and non-pyrogenic containers.
  • Made from durable 100% virgin material
  • Leakproof bottles and HDPE closures
  • Leakproof sealing with integrated lip seal feature
  • Sterile and non-pyrogenic, SAL 10⁻⁶
  • Low extractables
  • Transparency and reduced permeability
All containers are graduated, and feature a moulded hand grip for improved ergonomics.
Polycarbonate containers are suitable for use in temperatures ranging from –130 to 121 °C. PET/PETG containers are suitable for use in temperatures ranging from –70 to 60 °C.
Certificeringer: US FDA 21CRF, USP Class VI compliant resin.
Emballering: Double polybag packed.