Over-the-Spill Station
Leverandør: Rubbermaid Commercial Products
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FG425100YEL FG425200YEL FG425400YEL
129-1831EA 1990 DKK
129-1831 121-0199 121-0202
Over-the-Spill Station
Spild materialer
Dispensing station mounts on flat surfaces and has pads that soak up water and oil quickly and completely for an effective way to deal with spills.
  • Slip-resistant pads quickly cover spills and alert pedestrians
  • Each yellow pad is coded with universal caution symbol and 'Caution Wet Floor' message in English and Spanish
  • Melt-blown polypropylene pads absorb water and oil
The Rubbermaid Commercial over-the-spill station is a dispensing station that mounts on flat surfaces and includes pad dispenser, 22 large pads, and fasteners.
Over-the-Spill® Tablet:More Product Information