HYGEN™ Disposable Microfibre Pads
Leverandør: Rubbermaid Commercial Products
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2136051 2135887 2135889 2135890
RUBB2136051EA 3220 DKK
RUBB2136051 RUBB2135887 RUBB2135889 RUBB2135890
HYGEN™ Disposable Microfibre Pads
Mopper
Reduce cross-contamination with disposable microfibre pads.
  • Removes 99,7% or more of tested viruses and bacteria with water only
  • Reduce cross-contamination with a new single-use pad or cloth for each task
  • Facilitate colour-coding implementation by assigning a coloured pad to a specific area
  • Built-in zig-zag scrubbers effectively remove dirt and debris
  • Compatible with the common disinfectants Quad, Chlorine bleach, and Hydrogen Peroxide
  • Pads are ideal for use with the PULSE™ microfibre cleaning system
Rubbermaid Commercial HYGEN™ disposable microfibreMore Product Information