Chart Recorder Options for ULT Freezers
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
332166G01 332166G02 332791G01 332791G02
HERA332166G01EA 14220 DKK
HERA332166G01 HERA332166G02 HERA332791G01 HERA332791G02
Chart Recorder Options for ULT Freezers
Frysere Ultra lav temperatur frysere
Chart recorder options for ultra-low temperature chest freezers.
  • Ink or inkless, physical record keeping for temperature tracking
Chart recorders provide an uneditable, 21 CFR Part 11 compliant, historical temperature recording for documentation and tracking.
Bestillingsoplysninger: Add to ordering configuration with ULT chest freezer.