Backup Systems for ULT Freezers
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
332353G01 332353G02 332353G05 332353G06
HERA332353G01EA 27700 DKK
HERA332353G01 HERA332353G02 HERA332353G05 HERA332353G06
Backup Systems for ULT Freezers
Frysere Ultra lav temperatur frysere
CO₂ and LN₂ backup system options for ultra-low temperature chest freezers.
  • Factory installed, operates on independent backup probe
Sample security is paramount in cold storage and backup systems help ensure continued protection in the event of a refrigeration system issue with cold storage equipment. LN₂ and CO₂ backup options help maintain temperatures below −60 °C until a unit's primary refrigeration system returns to operation.
Bestillingsoplysninger: Add to ordering configuration with ULT chest freezer.