PermaSleeve® Heat-Shrink Labels for M710 and BMP71
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
174544 174541 174512 174615 174583 174527 174560 174545 174517 174511 174579 174654 174614 174518 174523 174610 174507 174524 174528 174559 174611 174632 174516 174564 174540 174563 174653 174628 174627 174655
BRDY174544EA 745 DKK
BRDY174544 BRDY174541 BRDY174512 BRDY174615 BRDY174583 BRDY174527 BRDY174560 BRDY174545 BRDY174517 BRDY174511 BRDY174579 BRDY174654 BRDY174614 BRDY174518 BRDY174523 BRDY174610 BRDY174507 BRDY174524 BRDY174528 BRDY174559 BRDY174611 BRDY174632 BRDY174516 BRDY174564 BRDY174540 BRDY174563 BRDY174653 BRDY174628 BRDY174627 BRDY174655
PermaSleeve® Heat-Shrink Labels for M710 and BMP71
Etiketter
PermaSleeve® PS Heat-Shrink Polyolefin (B-342) wire marking sleeves combine speed, ease of use, and permanence to create long-lasting, high-quality identification.
  • 3:1 heat shrinkable sleeve fits snugly around wires for maximum insulation protection and permanent identification
  • Fade-resistant and smudge-proof for superior aesthetic appearance
  • Cold resistant and flame-retardant with an operating temperature of –55 to +135 °C (–67 to +275 °F)
  • Matte finish
Applications include electrical labelling and wire and cable labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use Part Number M7-R4300.
Certificeringer: Meets SAE-AMS-DTL-23053/5C (Class 1), SAE-AS-81531, MIL-STD-202. RoHS compliant. UL recognised.