Easy-Tear Stopper Bags, with Steam Indicator, 1073B Tyvek/HDPE
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SBMGETHS0813 SBMGETHS1318 SBMGETHS1620 SBMGETHS2026 SBMGETHS2635 SBMGETHS3947
KACCSBMGETHS0813EA 13360 DKK
KACCSBMGETHS0813 KACCSBMGETHS1318 KACCSBMGETHS1620 KACCSBMGETHS2026 KACCSBMGETHS2635 KACCSBMGETHS3947
Easy-Tear Stopper Bags, with Steam Indicator, 1073B Tyvek/HDPE
Poser Steriliseringsposer
Easy-Tear Tyvek bags from Keystone Cleanroom Products are designed to satisfy the most stringent requirements in the sterilisation of critical pharmaceutical equipment and components.
  • Easy-Tear Feature for easier opening and sealing
  • No sterile cutting instruments required to open the bag
  • Low particle generation when opening via the Easy-Tear
  • Manufactured and packaged in a certified ISO 6 cleanroom
  • Exceptional microbial barrier
  • Also compatible with gamma irradiation, V₂H₂O and EtO sterilisation
Delivers outstanding performance in terms of cleanliness, material and seal strength, as well as excellent microbial barrier properties.
Leveringsinformation: Certificates of conformance sent with all shipments.
Emballering: Easy tear stopper bags are cleanroom packed in sealed, labeled poly bags with the Lot number and Date of manufacture.