Eppendorf twin.tec® Trace PCR Plates, 96 Wells
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0030129.768 0030129.776 0030129.784 0030129.822
EPPE0030129.768EA 1160 DKK
EPPE0030129.768 EPPE0030129.776 EPPE0030129.784 EPPE0030129.822
Eppendorf twin.tec® Trace PCR Plates, 96 Wells
Mikroplader PCR plader
The twin.tec® Trace PCR Plates have improved ergonomics and traceability. These microtiter plates are the perfect choice for high-throughput applications, automated workflows, and standardised processes.
  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • A new innovative optical guiding grid for quick well-orientation simplifies your manual and semi-automated liquid handling steps
  • Fuchsia and crystal blue colours support better visibility and contrast
  • Customized barcode versions are available
The new twin.tec Trace plates are comparable in terms of physical material, specifications and superior reproducibility to the well-established twin.tec plates.
Available in transparent and 2 fresh colours twin.tec Trace plates are ready to deliver maximum performance and reliability for your most demanding workflows.