Sterilisation Wraps, 2-Ply SMS, One-Step Heavy Duty
Leverandør: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KCRP1-300-0808 KCRP1-300-0909 KCRP1-300-0912 KCRP1-300-1212 KCRP1-300-1515 KCRP1-300-1818 KCRP1-300-2020 KCRP1-300-2424 KCRP1-300-3030 KCRP1-300-3636 KCRP1-300-4040 KCRP1-300-4545 KCRP1-300-4548 KCRP1-300-4848 KCRP1-300-5454 KCRP1-300-5472 KCRP1-300-5490 KCRP1-300-6060 KCRP1-300-6080
KACCKCRP1-300-0808EA 3290 DKK
KACCKCRP1-300-0808 KACCKCRP1-300-0909 KACCKCRP1-300-0912 KACCKCRP1-300-1212 KACCKCRP1-300-1515 KACCKCRP1-300-1818 KACCKCRP1-300-2020 KACCKCRP1-300-2424 KACCKCRP1-300-3030 KACCKCRP1-300-3636 KACCKCRP1-300-4040 KACCKCRP1-300-4545 KACCKCRP1-300-4548 KACCKCRP1-300-4848 KACCKCRP1-300-5454 KACCKCRP1-300-5472 KACCKCRP1-300-5490 KACCKCRP1-300-6060 KACCKCRP1-300-6080
Sterilisation Wraps, 2-Ply SMS, One-Step Heavy Duty
Steriliseringsindpakning
One-Step super duty SMS 2-ply wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications.
  • Manufactured and packaged in a certified ISO 8 Cleanroom
  • 2-ply
  • Clean - no cellulose
  • Strong, rip and tear resistant 88 gsm SMS material
  • An extensive range of sizes available means less cutting
  • Hydrophobic barrier allows sterilisation gases, but prevents liquid and microbial penetration
  • Gram weight: 50 (each layer)
For use in steam, ethylene oxide and hydrogen peroxide sterilisation processes.
Leveringsinformation: Certificate of Conformance sent with each shipment.
Emballering: Sub packed into sealed labeled poly bags with lot number and expiration date for complete traceability.