Cole-Parmer® WBU-200 Series Unheated Water Baths
Leverandør: Cole-Parmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
16101-88 16101-89 16104-45 16104-53 16112-01
TECN16101-88EA 6870 DKK
TECN16101-88 TECN16101-89 TECN16104-45 TECN16104-53 TECN16112-01
Cole-Parmer® WBU-200 Series Unheated Water Baths
Bade Vandbade, uden omrøring
Corrosion resistant baths with stainless steel inners for easy cleaning.
  • Constructed of durable stainless steel
  • Rugged splash-proof case
  • Integrated carrying handle for added safety
  • Material: Stainless steel
Designed to be used with the Cole-Parmer IC-300 series immersion circulators (sold separately), these water baths incorporate carrying handles for added safety. All water baths have stoved enamelled steel outer cases and are supplied with bridge mounting plate to hold the thermoregulator.