CRAIG GTX F2A ESD Firefighting Boots, Lace-Up
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
89641/35 89641/36 89641/37
ELTN89641/35EA 1790 DKK
ELTN89641/35 ELTN89641/36 ELTN89641/37
CRAIG GTX F2A ESD Firefighting Boots, Lace-Up
Sko og støvler
Firefighting boot, CRAIG GTX, in black finish.
  • Fire-resistant cowhide leather material
  • GORE-TEX performance comfort footwear
  • Full, padded bellows tongue
  • Full-length inlay sole FIREFIGHTERS Level 2
  • Anti-static soft fleece insole
  • Steel midsole
  • Rubber/PU tread sole POWERGRIP
  • TPU scuff cap
  • Steel toe cap
For use in outdoor areas with particularly high safety requirements, preferably for fire service operations. For areas with severe effects of heat, areas where there is a risk of chainsaw cuts, or areas where there is a risk of electrostatic discharge (More Product Information