STANTON S3 Safety Boots, Slip-On
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8631/39 8631/40 8631/41 8631/42 8631/43 8631/44 8631/45 8631/46 8631/47 8631/48 8631/49 8631/50
ELTN8631/39EA 895 DKK
ELTN8631/39 ELTN8631/40 ELTN8631/41 ELTN8631/42 ELTN8631/43 ELTN8631/44 ELTN8631/45 ELTN8631/46 ELTN8631/47 ELTN8631/48 ELTN8631/49 ELTN8631/50
STANTON S3 Safety Boots, Slip-On
Sko og støvler
Safety slip boot, STANTON, in silver/black finish.
  • Fire-resistant/aluminium-coated, heat-resistant cowhide leather
  • Leather lining
  • Full-length inlay sole, aluminium-coated
  • Non-metallic, penetration-resistant midsole
  • Rubber tread sole ZERMATT
  • Steel toe cap
For indoor and outdoor use, in areas where exposure to moisture is expected (S2) or areas where there is a risk of penetration from pointed and sharp objects (S3). For use in hot zones where high demands are placed on the sole for heat resistance, such as foundries, welding works etc. Specifically for areas where there is a risk of molten iron splashes or areas where there is a risk of molten aluminium splashes.
Certificeringer: EN ISO 20349-1 S3 HRO/Fe/Al/HI3/SRA, form C.